Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265841.1 | NP_001252770.1 | 551 | 153 | ||
GenBank mRNA | JU472820.1 | - | 628 | 153 | ||
JV636283.1 | - | 551 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |