Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015112692.2 | XP_014968178.1 | 1071 | 172 | ||
XM_015112694.2 | XP_014968180.1 | 1011 | 112 | |||
GenBank mRNA | JU325245.1 | - | 809 | 112 | ||
JU472254.1 | - | 1459 | 112 | |||
JV636321.1 | - | 2412 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |