Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IRX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001087101.4 | XP_001087101.3 | 2048 | 138 | ||
XM_028840930.1 | XP_028696763.1 | 1862 | 138 | |||
GenBank mRNA | JV723261.1 | - | 759 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |