Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266727.2 | NP_001253656.1 | 91 | 89 | ||
GenBank mRNA | JU323087.1 | - | 122 | 89 | ||
JU477308.1 | - | 91 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |