Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RILP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001086142.4 | XP_001086142.1 | 1085 | 108 | ||
XM_028835853.1 | XP_028691686.1 | 1079 | 108 | |||
XM_028835854.1 | XP_028691687.1 | 1085 | 108 | |||
GenBank mRNA | JV662885.1 | - | 128 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |