Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001085445.3 | XP_001085445.1 | 407 | 105 | ||
XM_015114062.2 | XP_014969548.1 | 652 | 105 | |||
XM_015114063.2 | XP_014969549.1 | 653 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |