Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: METTL21A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194117.3 | NP_001181046.1 | 259 | 65 | ||
XM_002799030.3 | XP_002799076.1 | 563 | 65 | |||
XM_002799031.3 | XP_002799077.1 | 387 | 65 | |||
XM_015110807.2 | XP_014966293.1 | 256 | 65 | |||
XM_028830557.1 | XP_028686390.1 | 565 | 65 | |||
XM_028830558.1 | XP_028686391.1 | 565 | 65 | |||
XM_028830559.1 | XP_028686392.1 | 564 | 65 | |||
XM_028830560.1 | XP_028686393.1 | 556 | 65 | |||
XM_028830561.1 | XP_028686394.1 | 387 | 65 | |||
XM_028830562.1 | XP_028686395.1 | 299 | 65 | |||
GenBank mRNA | JU321711.1 | - | 324 | 65 | ||
JU471680.1 | - | 279 | 65 | |||
JV045037.1 | - | 320 | 65 | |||
JV458095.1 | - | 237 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |