Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROSER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001082541.4 | XP_001082541.3 | 415 | 129 | ||
XM_028825985.1 | XP_028681818.1 | 1141 | 129 | |||
XM_028825986.1 | XP_028681819.1 | 448 | 129 | |||
XM_028825987.1 | XP_028681820.1 | 427 | 129 | |||
XM_028825988.1 | XP_028681821.1 | 1597 | 129 | |||
GenBank mRNA | JV647369.1 | - | 35 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |