Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM71D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194896.2 | NP_001181825.1 | 359 | 73 | ||
XM_015143939.2 | XP_014999425.2 | 941 | 73 | |||
XM_015143940.2 | XP_014999426.2 | 859 | 73 | |||
XM_015143941.2 | XP_014999427.2 | 940 | 73 | |||
XM_015143942.2 | XP_014999428.1 | 934 | 73 | |||
XM_015143944.2 | XP_014999430.2 | 280 | 73 | |||
XM_028850461.1 | XP_028706294.1 | 934 | 73 | |||
XM_028850462.1 | XP_028706295.1 | 934 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |