Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIMKLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261229.1 | NP_001248158.1 | 186 | 61 | ||
GenBank mRNA | JU334092.1 | - | 186 | 61 | ||
JU334093.1 | - | 186 | 61 | |||
JV613938.1 | - | 238 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |