Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC9C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194704.3 | NP_001181633.1 | 1099 | 95 | ||
GenBank mRNA | JU320168.1 | - | 760 | 95 | ||
JU472265.1 | - | 786 | 95 | |||
JV046801.1 | - | 726 | 95 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |