Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCTE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028846951.1 | XP_028702784.1 | 457 | 165 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |