Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001086077.4 | XP_001086077.1 | 481 | 106 | ||
GenBank mRNA | JU492878.1 | - | 447 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |