Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015114578.2 | XP_014970064.2 | 404 | 181 | ||
XM_015114580.2 | XP_014970066.2 | 404 | 181 | |||
XM_028833537.1 | XP_028689370.1 | 404 | 181 | |||
XM_028833538.1 | XP_028689371.1 | 404 | 181 | |||
XM_028833539.1 | XP_028689372.1 | 404 | 181 | |||
XM_028833540.1 | XP_028689373.1 | 404 | 181 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |