Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GOLT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001098173.4 | XP_001098173.1 | 208 | 136 | ||
XM_028852146.1 | XP_028707979.1 | 208 | 136 | |||
XM_028852149.1 | XP_028707982.1 | 208 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |