Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAPEPLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001086505.4 | XP_001086505.3 | 1644 | 115 | ||
XM_015134570.2 | XP_014990056.1 | 1483 | 115 | |||
XM_015134571.2 | XP_014990057.1 | 1151 | 115 | |||
XM_015134572.2 | XP_014990058.1 | 1166 | 115 | |||
XM_015134573.2 | XP_014990059.1 | 1258 | 115 | |||
XM_015134574.2 | XP_014990060.1 | 1184 | 115 | |||
XM_028846142.1 | XP_028701975.1 | 1083 | 115 | |||
GenBank mRNA | JU321104.1 | - | 1375 | 115 | ||
JU471371.1 | - | 1130 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |