Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015145909.2 | XP_015001395.1 | 491 | 136 | ||
XM_028852880.1 | XP_028708713.1 | 553 | 136 | |||
XM_028852882.1 | XP_028708715.1 | 540 | 136 | |||
GenBank mRNA | JU334491.1 | - | 450 | 136 | ||
JV529888.1 | - | 334 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |