Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLDN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JV044109.1 | - | 1165 | 127 | ||
JV635963.1 | - | 1099 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta |