Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PNOC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261798.1 | NP_001248727.1 | 583 | 76 | ||
XM_015145034.2 | XP_015000520.2 | 877 | 76 | |||
XM_015145035.2 | XP_015000521.2 | 602 | 76 | |||
GenBank mRNA | JU335036.1 | - | 583 | 76 | ||
JV548141.1 | - | 659 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta Inventoried |