Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195661.1 | NP_001182590.1 | 96 | 163 | ||
XM_015132415.2 | XP_014987901.1 | 1979 | 166 | |||
XM_015132416.2 | XP_014987902.1 | 162 | 166 | |||
XM_028843269.1 | XP_028699102.1 | 1664 | 163 | |||
GenBank mRNA | EG703357.1 | - | 101 | 163 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |