Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRDX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265826.2 | NP_001252755.1 | 374 | 118 | ||
GenBank mRNA | JU326514.1 | - | 189 | 118 | ||
JU479186.1 | - | 301 | 118 | |||
JV600249.1 | - | 374 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |