Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114113.1 | NP_001107585.1 | 2016 | 120 | ||
GenBank mRNA | EU340328.1 | - | 2016 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |