Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRYBB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122900.3 | NP_001116372.2 | 270 | 60 | ||
XM_028827337.1 | XP_028683170.1 | 2633 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Macaca mulatta |