Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193867.4 | NP_001180796.2 | 41 | 53 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |