Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RCCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193932.2 | NP_001180861.2 | 1015 | 87 | ||
GenBank mRNA | JU325061.1 | - | 872 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |