Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSKMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194712.2 | NP_001181641.1 | 320 | 92 | ||
XM_015113757.2 | XP_014969243.1 | 333 | 92 | |||
GenBank mRNA | JU472326.1 | - | 301 | 92 | ||
JV566752.1 | - | 313 | 92 | |||
JV635847.1 | - | 315 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |