Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC25A51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194642.2 | NP_001181571.1 | 227 | 102 | ||
GenBank mRNA | JU320769.1 | - | 381 | 102 | ||
JU477815.1 | - | 350 | 102 | |||
JU477816.1 | - | 181 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta |