Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ING1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265941.1 | NP_001252870.1 | 211 | 59 | ||
GenBank mRNA | JU472626.1 | - | 281 | 59 | ||
JV046977.1 | - | 211 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |