Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266612.1 | NP_001253541.1 | 366 | 90 | ||
XM_015150399.2 | XP_015005885.1 | 349 | 90 | |||
XM_015150400.2 | XP_015005886.1 | 559 | 90 | |||
GenBank mRNA | JU333220.1 | - | 465 | 90 | ||
JU474539.1 | - | 469 | 90 | |||
JV046339.1 | - | 366 | 90 | |||
JV573333.1 | - | 456 | 90 | |||
JV639477.1 | - | 488 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Macaca mulatta |