Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266883.2 | NP_001253812.1 | 1722 | 60 | ||
GenBank mRNA | JU322562.1 | - | 1542 | 60 | ||
JU473397.1 | - | 1714 | 60 | |||
JV043714.1 | - | 1726 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta |