Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Napa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080585.1 | NP_542152.1 | 7 - 8 | 607 | 84 | |
GenBank mRNA | BC063156.1 | - | 7 - 8 | 666 | 6921187 | 84 |
FQ213217.1 | - | 7 - 8 | 611 | 84 | ||
FQ216375.1 | - | 7 - 8 | 925 | 84 | ||
FQ234471.1 | - | 1 - 2 | 70 | 84 | ||
X89968.1 | - | 7 - 8 | 607 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |