Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053732.1 | NP_446184.1 | 2 - 3 | 1658 | 66 | |
XM_008767531.1 | XP_008765753.1 | 2 - 3 | 3213 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |