Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhcr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022389.2 | NP_071784.1 | 8 - 9 | 1397 | 62 | |
XM_006230892.2 | XP_006230954.1 | 7 - 8 | 1145 | 62 | ||
XM_006230893.2 | XP_006230955.1 | 7 - 8 | 1024 | 62 | ||
XM_006230894.2 | XP_006230956.1 | 8 - 9 | 1044 | 62 | ||
XM_008760170.1 | XP_008758392.1 | 8 - 9 | 2131 | 62 | ||
XM_008760171.1 | XP_008758393.1 | 7 - 8 | 1792 | 62 | ||
XM_008760172.1 | XP_008758394.1 | 8 - 9 | 1691 | 62 | ||
XM_008760173.1 | XP_008758395.1 | 7 - 8 | 1671 | 62 | ||
GenBank mRNA | AB016800.1 | - | 6 - 7 | 1088 | 62 | |
AF071500.1 | - | 7 - 8 | 1000 | 62 | ||
AF272393.1 | - | 8 - 9 | 1397 | 62 | ||
BC081688.1 | - | 6 - 7 | 1088 | 7109221 | 62 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |