Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dlk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053744.1 | NP_446196.1 | 205 | 74 | ||
XM_008764917.1 | XP_008763139.1 | 261 | 74 | |||
GenBank mRNA | BC167752.1 | - | 225 | 8365650 | 74 | |
D84336.1 | - | 205 | 74 | |||
U25680.1 | - | 70 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Rattus norvegicus 1700 Inventoried Probe spans exons |