Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpv5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053787.2 | NP_446239.2 | 3 - 4 | 503 | 81 | |
GenBank mRNA | AB032019.1 | - | 3 - 4 | 503 | 81 | |
AF209196.1 | - | 454 | 81 | |||
AY762624.1 | - | 3 - 4 | 338 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Ion Channel Detect Genomic DNA Rattus norvegicus Probe spans exons |