Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3gat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054003.1 | NP_446455.1 | 1 - 2 | 103 | 62 | |
XM_006242733.2 | XP_006242795.1 | 1 - 2 | 375 | 62 | ||
XM_006242734.2 | XP_006242796.1 | 1 - 2 | 348 | 62 | ||
GenBank mRNA | D88035.1 | - | 1 - 2 | 103 | 62 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Rattus norvegicus Inventoried Probe spans exons |