Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr7a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_134407.1 | NP_599234.1 | 1 - 2 | 292 | 114 | |
GenBank mRNA | AB037424.1 | - | 1 - 2 | 292 | 114 | |
AF503514.1 | - | 1 - 2 | 330 | 114 | ||
AJ271883.1 | - | 1 - 2 | 295 | 114 | ||
BC061816.1 | - | 1 - 2 | 295 | 5598724 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus 1700 Inventoried Probe spans exons |