Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Obp1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138903.1 | NP_620258.1 | 522 | 73 | ||
GenBank mRNA | J03093.1 | - | 522 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Inventoried Probe spans exons |