Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zwint | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_147138.2 | NP_671479.1 | 6 - 7 | 704 | 81 | |
GenBank mRNA | AF439397.1 | - | 6 - 7 | 686 | 81 | |
AF532776.1 | - | 6 - 7 | 704 | 81 | ||
BC063144.1 | - | 6 - 7 | 696 | 6921645 | 81 | |
EU000468.1 | - | 6 - 7 | 672 | 81 | ||
FQ228814.1 | - | 4 - 5 | 438 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Rattus norvegicus 1700 Probe spans exons |