Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gata2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033442.1 | NP_254277.1 | 5 - 6 | 1466 | 73 | |
GenBank mRNA | AB102789.1 | - | 3 - 4 | 1023 | 73 | |
AB102790.1 | - | 3 - 4 | 1023 | 73 | ||
AF345897.1 | - | 3 - 4 | 1023 | 73 | ||
AY032734.1 | - | 5 - 6 | 1466 | 73 | ||
BC061745.1 | - | 4 - 5 | 1273 | 5599740 | 73 | |
FQ212304.1 | - | 1347 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Rattus norvegicus Probe spans exons |