Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mapk8ip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053777.1 | NP_446229.1 | 5 - 6 | 1601 | 55 | |
GenBank mRNA | AF108959.1 | - | 4 - 5 | 1539 | 55 | |
AF109772.1 | - | 4 - 5 | 1448 | 55 | ||
AF109773.1 | - | 4 - 5 | 1511 | 55 | ||
AF109774.1 | - | 5 - 6 | 1601 | 55 | ||
DQ377223.1 | - | 4 - 5 | 1411 | 55 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |