Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053931.3 | NP_446383.3 | 3 - 4 | 530 | 79 | |
GenBank mRNA | AB027143.1 | - | 3 - 4 | 530 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Rattus norvegicus Probe spans exons |