Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Des | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022531.1 | NP_071976.1 | 7 - 8 | 1363 | 70 | |
GenBank mRNA | BC061872.1 | - | 7 - 8 | 1356 | 5598951 | 70 |
FQ216012.1 | - | 3 - 4 | 329 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |