Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cspg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031022.1 | NP_112284.1 | 4 - 5 | 4357 | 65 | |
GenBank mRNA | X56541.2 | - | 4 - 5 | 4357 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Rattus norvegicus Probe spans exons |