Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012520.2 | NP_036652.1 | 2 - 3 | 369 | 68 | |
GenBank mRNA | BC081853.1 | - | 2 - 3 | 266 | 7109889 | 68 |
FM043772.1 | - | 2 - 3 | 369 | 68 | ||
M11670.1 | - | 2 - 3 | 320 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Rattus norvegicus Probe spans exons |