Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcrtr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013064.1 | NP_037196.1 | 4 - 5 | 945 | 81 | |
GenBank mRNA | AF041244.1 | - | 4 - 5 | 945 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 G-protein coupled receptor Rattus norvegicus Probe spans exons |