Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gys2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF346902.1 | - | 2 - 3 | 308 | 66 | |
J05446.1 | - | 2 - 3 | 353 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Rattus norvegicus Probe spans exons |