Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r2d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144746.2 | NP_653347.2 | 6 - 7 | 762 | 91 | |
GenBank mRNA | AF180350.1 | - | 6 - 7 | 766 | 91 | |
BC081720.1 | - | 6 - 7 | 762 | 7112646 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Protein Phosphatase Mammalian Gene Collection Rattus norvegicus Probe spans exons |