Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor1aip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145092.2 | NP_659560.2 | 2 - 3 | 749 | 121 | |
XM_006250027.2 | XP_006250089.1 | 2 - 3 | 294 | 121 | ||
XM_006250028.2 | XP_006250090.1 | 2 - 3 | 880 | 121 | ||
GenBank mRNA | BC086987.1 | - | 2 - 3 | 749 | 7129718 | 121 |
U19614.1 | - | 2 - 3 | 619 | 121 | ||
U20286.1 | - | 2 - 3 | 206 | 121 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Rattus norvegicus Probe spans exons |