Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Men1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019208.3 | NP_062081.1 | 5 - 6 | 1312 | 101 | |
XM_006230769.2 | XP_006230831.1 | 6 - 7 | 1030 | 101 | ||
XM_006230770.2 | XP_006230832.1 | 6 - 7 | 1121 | 101 | ||
XM_006230771.2 | XP_006230833.1 | 6 - 7 | 1031 | 101 | ||
XM_006230772.2 | XP_006230834.1 | 6 - 7 | 1089 | 101 | ||
GenBank mRNA | AB023400.1 | - | 5 - 6 | 1379 | 101 | |
AF130369.1 | - | 5 - 6 | 1287 | 101 | ||
AF130370.1 | - | 5 - 6 | 932 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Rattus norvegicus Probe spans exons |